5-Pyrimidineacetic acid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD105337-1GEA 247 EUR
BLDPBD105337-1G BLDPBD105337-250MG BLDPBD105337-5G
5-Pyrimidineacetic acid 97%
5-Pyrimidineacetic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C6H6N2O2
MW: 138,1240 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09608122
CAS-nummer: 5267-07-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR