Dimethyl-4,5-imidazoledicarboxylate 98%

BLDPBD10569-10GEA 37 EUR
BLDPBD10569-10G BLDPBD10569-500G BLDPBD10569-1G BLDPBD10569-25G BLDPBD10569-100G BLDPBD10569-250MG BLDPBD10569-5G
Dimethyl-4,5-imidazoledicarboxylate 98%
Dimethyl-4,5-imidazoledicarboxylate

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₈N₂O₄
MW: 184,1500 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00051338
CAS-nummer: 3304-70-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR