m-Terphenyl 99.5%

Leverancier: BLD PHARMATECH GMBH

BLDPBD105814-25GEA 248 EUR
BLDPBD105814-25G BLDPBD105814-5G BLDPBD105814-1G
m-Terphenyl 99.5%
m-Terphenyl

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₈H₁₄
MW: 230,31 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003059
CAS-nummer: 92-06-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR