4-Bromo-2-furoic acid 97%

BLDPBD106995-250MGEA 65 EUR
BLDPBD106995-250MG BLDPBD106995-1G BLDPBD106995-5G
4-Bromo-2-furoic acid 97%
4-Bromo-2-furoic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₅H₃BrO₃
MW: 190,9800 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00092494
CAS-nummer: 3439-02-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR