(R)-(-)-2-Octanol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD10769-100GEA 555 EUR
BLDPBD10769-100G BLDPBD10769-25G BLDPBD10769-10G BLDPBD10769-5G BLDPBD10769-1G
(R)-(-)-2-Octanol 98%
(R)-(-)-2-Octanol

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₁₈O
MW: 130,23 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00064284
CAS-nummer: 5978-70-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR