6,6'-Dimethyl-2,2'-bipyridyl 98%

BLDPBD107780-10GEA 84 EUR
BLDPBD107780-10G BLDPBD107780-5G BLDPBD107780-1G BLDPBD107780-100G BLDPBD107780-250MG BLDPBD107780-25G
6,6'-Dimethyl-2,2'-bipyridyl 98%
6,6'-Dimethyl-2,2'-bipyridyl

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₂H₁₂N₂
MW: 184,2371 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00059776
CAS-nummer: 4411-80-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR