2-Amino-4-(trifluoromethyl)benzonitrile 98%

BLDPBD10895-25GEA 153 EUR
BLDPBD10895-25G BLDPBD10895-100G BLDPBD10895-5G BLDPBD10895-10G BLDPBD10895-1G
2-Amino-4-(trifluoromethyl)benzonitrile 98%
2-Amino-4-(trifluoromethyl)benzonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₅F₃N₂
MW: 186,1339 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04974124
CAS-nummer: 1483-54-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR