3',4'-Dichloroacetophenone 98%

BLDPBD10942-10GEA 10 EUR
BLDPBD10942-10G BLDPBD10942-100G BLDPBD10942-500G BLDPBD10942-5G BLDPBD10942-25G
3',4'-Dichloroacetophenone 98%
3',4'-Dichloroacetophenone

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₆Cl₂O
MW: 189,04 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00000553
CAS-nummer: 2642-63-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR