p-Tolyl isocyanate 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD11076-5GEA 6 EUR
BLDPBD11076-5G BLDPBD11076-25G
p-Tolyl isocyanate 98%
p-Tolyl isocyanate

Storage condition: Inert atmosphere, 2-8°C

Formule: C₈H₇NO
MW: 133,15 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00002029
CAS-nummer: 622-58-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR