6-Nitroindoline 97%

BLDPBD11136-100GEA 334 EUR
BLDPBD11136-100G BLDPBD11136-5G BLDPBD11136-1G BLDPBD11136-25G
6-Nitroindoline 97%
6-Nitroindoline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₈H₈N₂O₂
MW: 164,16 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00005710
CAS-nummer: 19727-83-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR