Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD11166-5GEA 7 EUR
BLDPBD11166-5G BLDPBD11166-100G BLDPBD11166-500G BLDPBD11166-25G BLDPBD11166-10G
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 97%
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₉ClN₂O₂S
MW: 232,69 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00006085
CAS-nummer: 5909-24-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR