4,4-Dimethoxybutanone 90%

BLDPBD112288-25GEA 7 EUR
BLDPBD112288-25G BLDPBD112288-10G
4,4-Dimethoxybutanone 90%
4,4-Dimethoxybutanone

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₁₂O₃
MW: 132,16 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00008789
CAS-nummer: 5436-21-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR