Azelaic acid 98%

BLDPBD114904-25GEA 6 EUR
BLDPBD114904-25G BLDPBD114904-1000G BLDPBD114904-100G BLDPBD114904-500G
Azelaic acid 98%
Azelaic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₁₆O₄
MW: 188,22 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00004432
CAS-nummer: 123-99-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR