Ethyl-4-fluoro-3-nitrobenzoate 98%
|
|
BLDPBD11630-10GEA
22
EUR
BLDPBD11630-10G
BLDPBD11630-25G
BLDPBD11630-1G
BLDPBD11630-5G
BLDPBD11630-100G
Ethyl-4-fluoro-3-nitrobenzoate 98%
Ethyl-4-fluoro-3-nitrobenzoate
Storage condition: Sealed in dry, Room Temperature
Formule:
C₉H₈FNO₄ MW: 213,1600 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD03428515 CAS-nummer: 367-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...