(S)-(+)-O-Acetylmandelic acid 97%

BLDPBD11846-10GEA 7 EUR
BLDPBD11846-10G BLDPBD11846-500G BLDPBD11846-5G BLDPBD11846-25G BLDPBD11846-100G
(S)-(+)-O-Acetylmandelic acid 97%
(S)-(+)-O-Acetylmandelic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₀H₁₀O₄
MW: 194,19 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00064215
CAS-nummer: 7322-88-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR