(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 95%
|
|
BLDPBD1194-25GEA
24
EUR
BLDPBD1194-25G
BLDPBD1194-100G
BLDPBD1194-10G
BLDPBD1194-0.025G
BLDPBD1194-5G
BLDPBD1194-500G
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formule:
C₂₀H₁₉NO₆ MW: 369,4 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00237657 CAS-nummer: 121343-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...