Calcium hydrogen orthophosphate dihydrate 99%

Leverancier: BLD PHARMATECH GMBH

BLDPBD119683-0.5GEA 0 EUR
BLDPBD119683-0.5G BLDPBD119683-1000G BLDPBD119683-100G BLDPBD119683-500G
Calcium hydrogen orthophosphate dihydrate 99%
Calcium hydrogen orthophosphate dihydrate

Storage condition: Inert atmosphere, Room Temperature

Formule: HCaO₄P·2H₂O
MW: 172,09 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00149621
CAS-nummer: 7789-77-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR