(S)-Methyl-2-hydroxy-3-phenylpropanoate 98%

BLDPBD12059-100GEA 156 EUR
BLDPBD12059-100G BLDPBD12059-5G BLDPBD12059-500G BLDPBD12059-25G BLDPBD12059-10G BLDPBD12059-1G
(S)-Methyl-2-hydroxy-3-phenylpropanoate 98%
(S)-Methyl-2-hydroxy-3-phenylpropanoate

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₁₂O₃
MW: 180,2005 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00038224
CAS-nummer: 13673-95-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR