4-(Dimethylamino)-2-methoxybenzaldehyde 95%

BLDPBD12151-5GEA 231 EUR
BLDPBD12151-5G BLDPBD12151-1G BLDPBD12151-250MG
4-(Dimethylamino)-2-methoxybenzaldehyde 95%
4-(Dimethylamino)-2-methoxybenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formule: C₁₀H₁₃NO₂
MW: 179,22 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00151814
CAS-nummer: 84562-48-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR