Cyclobutylmethanamine hydrochloride 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD122614-250MGEA 9 EUR
BLDPBD122614-250MG BLDPBD122614-5G BLDPBD122614-100G BLDPBD122614-25G BLDPBD122614-1G
Cyclobutylmethanamine hydrochloride 97%
Cyclobutylmethanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₅H₁₂ClN
MW: 121,6100 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00034952
CAS-nummer: 5454-82-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR