N-Boc-O-methyl-L-threonine 96%

Leverancier: BLD PHARMATECH GMBH

BLDPBD12398-25GEA 121 EUR
BLDPBD12398-25G BLDPBD12398-1G BLDPBD12398-10G BLDPBD12398-100G BLDPBD12398-5G
N-Boc-O-methyl-L-threonine 96%
N-Boc-O-methyl-L-threonine

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₀H₁₉NO₅
MW: 233,3 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00076990
CAS-nummer: 48068-25-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR