Diethyl phthalate 99%

Leverancier: BLD PHARMATECH GMBH

BLDPBD124754-25GEA 6 EUR
BLDPBD124754-25G
Diethyl phthalate 99%
Diethyl phthalate

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₂H₁₄O₄
MW: 222,24 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00009111
CAS-nummer: 84-66-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR