Dimethyl pimelate 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD125456-10GEA 37 EUR
BLDPBD125456-10G BLDPBD125456-25G BLDPBD125456-100G BLDPBD125456-5G
Dimethyl pimelate 98%
Dimethyl pimelate

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₁₆O₄
MW: 188,22 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00008470
CAS-nummer: 1732-08-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR