Dimethyl sebacate 98%

BLDPBD125466-25GEA 6 EUR
BLDPBD125466-25G
Dimethyl sebacate 98%
Dimethyl sebacate

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₂H₂₂O₄
MW: 230,3 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00008472
CAS-nummer: 106-79-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR