di-Sodium L(+)-tartrate dihydrate 99+%

Leverancier: BLD PHARMATECH GMBH

BLDPBD126091-25GEA 11 EUR
BLDPBD126091-25G
di-Sodium L(+)-tartrate dihydrate 99+%
di-Sodium L(+)-tartrate dihydrate
Formule: NaOOC(CHOH)₂COONa·2H₂O
MW: 230,08 g/mol
MDL-nummer: MFCD00150035
CAS-nummer: 6106-24-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR