Isoquinoline-3-carboxylic acid 97%

BLDPBD12623-1GEA 16 EUR
BLDPBD12623-1G BLDPBD12623-10G BLDPBD12623-5G BLDPBD12623-25G BLDPBD12623-250MG
Isoquinoline-3-carboxylic acid 97%
Isoquinoline-3-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₁₀H₇NO₂
MW: 173,1681 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00075137
CAS-nummer: 6624-49-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR