1,3-Dihydroxynaphthalene 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD12743-100MGEA 6 EUR
BLDPBD12743-100MG BLDPBD12743-10G BLDPBD12743-0.025G BLDPBD12743-25G BLDPBD12743-100G BLDPBD12743-5G BLDPBD12743-1G BLDPBD12743-250MG
1,3-Dihydroxynaphthalene 98%
1,3-Dihydroxynaphthalene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₁₀H₈O₂
MW: 160,17 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003965
CAS-nummer: 132-86-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR