(4-Chlorophenyl)acetic acid ethyl ester 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD128859-10GEA 6 EUR
BLDPBD128859-10G BLDPBD128859-500G BLDPBD128859-100G BLDPBD128859-25G
(4-Chlorophenyl)acetic acid ethyl ester 95%
(4-Chlorophenyl)acetic acid ethyl ester

Storage condition: Sealed in dry, Room Temperature

Formule: (C₆H₄Cl)CH₂COOCH₂CH₃
MW: 198,65 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00018791
CAS-nummer: 14062-24-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR