Ethyl hippurate 95+%

Leverancier: BLD PHARMATECH GMBH

BLDPBD129354-1GEA 8 EUR
BLDPBD129354-1G BLDPBD129354-100G BLDPBD129354-5G BLDPBD129354-25G
Ethyl hippurate 95+%
Ethyl hippurate

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₁H₁₃NO₃
MW: 207,23 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00026890
CAS-nummer: 1499-53-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR