Ethyl linoleate 70%

Leverancier: BLD PHARMATECH GMBH

BLDPBD129400-0.025EA 0 EUR
BLDPBD129400-0.025 BLDPBD129400-25G BLDPBD129400-100G BLDPBD129400-500G
Ethyl linoleate 70%
Ethyl linoleate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₂₀H₃₆O₂
MW: 308,5 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00009535
CAS-nummer: 544-35-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR