6,7-Dimethoxyquinolin-4-ol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD13035-100GEA 188 EUR
BLDPBD13035-100G BLDPBD13035-1G BLDPBD13035-25G BLDPBD13035-5G BLDPBD13035-10G
6,7-Dimethoxyquinolin-4-ol 98%
6,7-Dimethoxyquinolin-4-ol

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₁H₁₁NO₃
MW: 205,2099 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09031937
CAS-nummer: 13425-93-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR