2-Fluoro-α-methyl-4-biphenylacetic acid 98%

Synoniemen: FrobenAnsaidAntadysFlurbiprofen

BLDPBD130642-25GEA 151 EUR
BLDPBD130642-25G BLDPBD130642-10G BLDPBD130642-1G BLDPBD130642-500G BLDPBD130642-5G BLDPBD130642-100G
2-Fluoro-α-methyl-4-biphenylacetic acid 98%
2-Fluoro-α-methyl-4-biphenylacetic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₅H₁₃FO₂
MW: 244,265 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00079303
CAS-nummer: 5104-49-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR