1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD13116-10GEA 34 EUR
BLDPBD13116-10G BLDPBD13116-25G BLDPBD13116-500G BLDPBD13116-5G BLDPBD13116-100G BLDPBD13116-1G
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 98%
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₁H₁₂O₂
MW: 176,22 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02179127
CAS-nummer: 1914-65-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR