(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 97%

BLDPBD132926-1GEA 7 EUR
BLDPBD132926-1G BLDPBD132926-10G BLDPBD132926-5G BLDPBD132926-25G
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 97%
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formule: C₁₃H₁₉NO₂·HCl
MW: 257,8 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD00153455
CAS-nummer: 3403-25-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR