1-Boc-piperidine-3-carboxylic acid 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD13459-5GEA 7 EUR
BLDPBD13459-5G BLDPBD13459-25G BLDPBD13459-500G BLDPBD13459-100G BLDPBD13459-10G
1-Boc-piperidine-3-carboxylic acid 95%
1-Boc-piperidine-3-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₁H₁₉NO₄
MW: 229,3 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00673775
CAS-nummer: 84358-12-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR