Lactobionic acid 97%
|
|
BLDPBD135770-5GEA
7
EUR
BLDPBD135770-5G
BLDPBD135770-1G
BLDPBD135770-100G
BLDPBD135770-500G
BLDPBD135770-25G
Lactobionic acid 97%
Lactobionic acid
Storage condition: Inert atmosphere, Room Temperature
Formule:
C₁₂H₂₂O₁₂ MW: 358,30 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00078147 CAS-nummer: 96-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...