2-Chloro-6-methylquinoline-3-carbaldehyde 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD13622-250MGEA 7 EUR
BLDPBD13622-250MG BLDPBD13622-1G
2-Chloro-6-methylquinoline-3-carbaldehyde 97%
2-Chloro-6-methylquinoline-3-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formule: C₁₁H₈ClNO
MW: 205,6400 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD01912773
CAS-nummer: 73568-27-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR