Methyl-3-methylsalicylate 98%

BLDPBD138295-1GEA 16 EUR
BLDPBD138295-1G BLDPBD138295-25G BLDPBD138295-5G
Methyl-3-methylsalicylate 98%
Methyl-3-methylsalicylate

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₁₀O₃
MW: 166,18 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00020051
CAS-nummer: 23287-26-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR