1,3,3-Trimethyl-2-methyleneindoline 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD14054-100GEA 61 EUR
BLDPBD14054-100G BLDPBD14054-500G BLDPBD14054-5G BLDPBD14054-25G
1,3,3-Trimethyl-2-methyleneindoline 98%
1,3,3-Trimethyl-2-methyleneindoline

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₂H₁₅N
MW: 173,26 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005813
CAS-nummer: 118-12-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR