Naphthacene 98%

BLDPBD143252-250MGEA 79 EUR
BLDPBD143252-250MG BLDPBD143252-5G BLDPBD143252-100MG BLDPBD143252-1G
Naphthacene 98%
Naphthacene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₁₈H₁₂
MW: 228,29 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003702
CAS-nummer: 92-24-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR