N-Boc-aniline 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD143556-25GEA 62 EUR
BLDPBD143556-25G BLDPBD143556-5G BLDPBD143556-500G BLDPBD143556-100G
N-Boc-aniline 95%
N-Boc-aniline

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₁H₁₅NO₂
MW: 193,2423 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00043723
CAS-nummer: 3422-01-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR