(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD14895-5GEA 15 EUR
BLDPBD14895-5G BLDPBD14895-100G BLDPBD14895-25G BLDPBD14895-1G BLDPBD14895-10G
(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine 95%
(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₃H₁₈FNO
MW: 223,29 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06658161
CAS-nummer: 105812-81-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR