2-(1H-Pyrazol-4-yl)pyridine 95%

BLDPBD149706-1GEA 61 EUR
BLDPBD149706-1G BLDPBD149706-5G BLDPBD149706-100MG BLDPBD149706-250MG
2-(1H-Pyrazol-4-yl)pyridine 95%
2-(1H-Pyrazol-4-yl)pyridine

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₇N₃
MW: 145,1613 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02579318
CAS-nummer: 439106-75-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR