Pyrimidine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD149827-1GEA 7 EUR
BLDPBD149827-1G BLDPBD149827-5G BLDPBD149827-25G
Pyrimidine 98%
Pyrimidine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₄H₄N₂
MW: 80,09 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00006059
CAS-nummer: 289-95-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR