Quinoxaline 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD150129-1000GEA 228 EUR
BLDPBD150129-1000G BLDPBD150129-100G BLDPBD150129-500G BLDPBD150129-5G BLDPBD150129-10G BLDPBD150129-25G
Quinoxaline 97%
Quinoxaline

Storage condition: Inert atmosphere, Room Temperature

Formule: C₈H₆N₂
MW: 130,15 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00006719
CAS-nummer: 91-19-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR