4-Benzyloxy-2-formylphenylboronic acid 95%
|
|
BLDPBD15333-250MGEA
17
EUR
BLDPBD15333-250MG
BLDPBD15333-5G
BLDPBD15333-1G
BLDPBD15333-25G
4-Benzyloxy-2-formylphenylboronic acid 95%
4-Benzyloxy-2-formylphenylboronic acid
Storage condition: Inert atmosphere, Room Temperature
Formule:
C₁₄H₁₃BO₄ MW: 256,0616 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD02179486 CAS-nummer: 139962-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...