Thieno[3,2-d]pyrimidin-4(3H)-one 98%
|
|
BLDPBD153867-25GEA
84
EUR
BLDPBD153867-25G
BLDPBD153867-100G
BLDPBD153867-5G
BLDPBD153867-1G
Thieno[3,2-d]pyrimidin-4(3H)-one 98%
Thieno[3,2-d]pyrimidin-4(3H)-one
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formule:
C₆H₄N₂OS MW: 152,17 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD03407527 CAS-nummer: 16234-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...