Tin(II) sulphate 99%
|
|
BLDPBD154172-25GEA
7
EUR
BLDPBD154172-25G
BLDPBD154172-0.025
Tin(II) sulphate 99%
Tin(II) sulphate
Storage condition: Inert atmosphere, Room Temperature
Formule:
O₄SSn MW: 214,77 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00011246 CAS-nummer: 7488-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...