3-(Boc-amino)-3-(2-fluorophenyl)propionic acid 98%

BLDPBD15513-5GEA 245 EUR
BLDPBD15513-5G BLDPBD15513-250MG BLDPBD15513-1G
3-(Boc-amino)-3-(2-fluorophenyl)propionic acid 98%
3-(Boc-amino)-3-(2-fluorophenyl)propionic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₄H₁₈FNO₄
MW: 283,30 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD02090698
CAS-nummer: 284493-56-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR