3-(Boc-amino)-3-phenylpropionic acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD15569-25GEA 166 EUR
BLDPBD15569-25G BLDPBD15569-100G BLDPBD15569-5G BLDPBD15569-1G
3-(Boc-amino)-3-phenylpropionic acid 98%
3-(Boc-amino)-3-phenylpropionic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₄H₁₉NO₄
MW: 265,31 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02090695
CAS-nummer: 14676-01-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR